(+)-Di-p-toluoyl-D-tartaric acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid
L13223-14
L13223-22
CAAAAL13223-06EA
93.77
CAD
CAAAAL13223-06
CAAAAL13223-14
CAAAAL13223-22
(+)-Di-p-toluoyl-D-tartaric acid 98%
(+)-Di-p-toluoyl-D-tartaric acid
Purity: 98%
CAS No.: 32634-68-7
Molecular Formula: C20H18O8
Formula Weight: 386.36
Melting Point: 169-171°
Beilstein Reference: 3225584
Formula:
C₂₀H₁₈O₈ MW: 386.36 g/mol Melting Pt: 169…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008552 CAS Number: 32634-68-7 EINECS: 251-132-2 |
Specification Test Results
Optical rotation | +139° (c=1 in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...