Benzoyl peroxide 97% (dry weight) wetted with ca. 25% water

Supplier: Thermo Scientific Chemicals
Danger

L13174-0B L13174-18 L13174-30
CAAAAL13174-0BEA 221.2 CAD
CAAAAL13174-0B CAAAAL13174-18 CAAAAL13174-30
Benzoyl peroxide 97% (dry weight) wetted with ca. 25% water
Benzoyl peroxide

Fieser: 1,196 4,122 5,182 6,160 7,89 12,157 18,122 20,130


Purity: 97% CAS No.: 94-36-0 Molecular Formula: C14H10O4 Formula Weight: 242.23 Beilstein Reference: 984320 Merck Reference: 14,1116 MDL No.: MFCD00003071

Formula: C₁₄H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00003071
CAS Number: 94-36-0
EINECS: 202-327-6
UN: 3104
ADR: "5.2",
Merck Index: 14,1116

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR