Acridine orange (hemizinc salt)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride
Fluorescent stain for proteins. An RNA polymerase inhibitor.
CAS No.: 10127-02-3
Molecular Formula: C17H19N3.HCl.ZnCl2
Formula Weight: 438.10
Beilstein Reference: 3734978
MDL No.: MFCD00081043
Formula:
C₁₇H₂₀ClN₃·HCl·½ZnCl₂ MW: 369.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00081043 CAS Number: 10127-02-3 EINECS: 233-353-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...