3-(3-Furyl)acrylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13143-03
L13143-06
L13143-14
CAAAAL13143-03EA
97.8
CAD
CAAAAL13143-03
CAAAAL13143-06
CAAAAL13143-14
3-(3-Furyl)acrylic acid 98%
3-(3-Furyl)acrylic acid
Purity: 98%
CAS No.: 39244-10-5
Molecular Formula: C7H6O3
Formula Weight: 138.12
Melting Point: 152-154°
Beilstein Reference: 1306450
Formula:
C₇H₆O₃ Melting Pt: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075074 CAS Number: 39244-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...