2(5H)-Furanone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Furan-2(5H)-one

L13142-03 L13142-06 L13142-14
CAAAAL13142-03EA 116.45 CAD
CAAAAL13142-03 CAAAAL13142-06 CAAAAL13142-14
2(5H)-Furanone 96%
2(5H)-Furanone

Purity: 98% CAS No.: 497-23-4 Molecular Formula: C4H4O2 Formula Weight: 84.07 Storage Temperature: 2°C to 8°C Flash Point: 101°(213°F) Refractive Index: 1.4690 Melting Point: 3-4° Boiling Point: 212-214° Beilstein Reference: 383585 MDL No.: MFCD00005376

Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 212…214 °C
Melting Pt: 3…4 °C
Density: 1.197
Flash Pt: 101 °C(213 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005376
CAS Number: 497-23-4
EINECS: 207-839-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR