2-Amino-4,4,4-trifluorobutyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-2-Amino-4,4,4-trifluorobutyric acid, 2-Amino-4,4,4-trifluorobutanoic acid
L13131-MC
L13131-ME
CAAAAL13131-MCEA
62.25
CAD
CAAAAL13131-MC
CAAAAL13131-ME
2-Amino-4,4,4-trifluorobutyric acid 97%
(±)-2-Amino-4,4,4-trifluorobutyric acid
Purity: 97%
CAS No.: 15959-93-0
Molecular Formula: C4H6F3NO2
Formula Weight: 157.09
Melting Point: ca 250° dec.
Beilstein Reference: 1769355
Formula:
C₄H₆F₃NO₂ MW: 157.09 g/mol Melting Pt: ∼ 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00041417 CAS Number: 15959-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...