(4-Nitrophenyl)-β-D-galactopyranoside hydrate 98+%
Supplier: Thermo Scientific Chemicals
L12911-03
L12911-06
CAAAAL12911-03EA
64.94
CAD
CAAAAL12911-03
CAAAAL12911-06
(4-Nitrophenyl)-β-D-galactopyranoside hydrate 98+%
(4-Nitrophenyl) β-D-galactopyranoside hydrate
Purity: 98+%
CAS No.: 200422-18-0
Molecular Formula: C12H15NO8.xH2O
Formula Weight: 319.27 (301.25anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 180-184°
Beilstein Reference: 92213
Optical Rotation: -84° (c=0.3 in water)
MDL No.: MFCD00150725
Formula:
C₁₂H₁₅NO₈·H₂O MW: 319.27 g/mol (301.25 anhy) Melting Pt: 180…184 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00150725 CAS Number: 200422-18-0 EINECS: 221-584-5 |
Specification Test Results
Optical rotation | -84° (c=0.3 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...