DL-α-Aminoadipic acid 98%
Supplier: Thermo Scientific Chemicals
L12717-03
L12717-06
CAAAAL12717-03EA
147.26
CAD
CAAAAL12717-03
CAAAAL12717-06
DL-α-Aminoadipic acid 98%
DL-α-Aminoadipic acid
Purity: 98%
CAS No.: 542-32-5
Molecular Formula: C6H11NO4
Formula Weight: 161.16
Melting Point: 196-198°
Beilstein Reference: 1773077
Merck Reference: 14,416
MDL No.: MFCD00063119
Formula:
C₆H₁₁NO₄ Melting Pt: 196…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063119 CAS Number: 542-32-5 EINECS: 208-809-2 Merck Index: 14,416 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...