1,4-Butanediol diglycidyl ether 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12632-09
L12632-18
CAAAAL12632-09EA
109.34
CAD
CAAAAL12632-09
CAAAAL12632-18
1,4-Butanediol diglycidyl ether 96%
1,4-Butanediol diglycidyl ether
Purity: 96%
CAS No.: 2425-79-8
Molecular Formula: C10H18O4
Formula Weight: 202.25
Flash Point: 140°(284°F)
Refractive Index: 1.4550
Boiling Point: 157-158°/11mm
Beilstein Reference: 115238
MDL No.: MFCD00005146
Formula:
C₁₀H₁₈O₄ Boiling Pt: 157…158 °C/11 mm Density: 1.049 Flash Pt: 140 °C(284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005146 CAS Number: 2425-79-8 EINECS: 219-371-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...