2,6-Dinitrobenzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12624-03
L12624-06
CAAAAL12624-03EA
143.46
CAD
CAAAAL12624-03
CAAAAL12624-06
2,6-Dinitrobenzonitrile 98+%
2,6-Dinitrobenzonitrile
Purity: 98+%
CAS No.: 35213-00-4
Molecular Formula: C7H3N3O4
Formula Weight: 193.12
Melting Point: 146-150°
Beilstein Reference: 2215516
UN No.: 3439
MDL No.: MFCD00041246
Formula:
C₇H₃N₃O₄ Melting Pt: 146…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041246 CAS Number: 35213-00-4 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...