(±)-3-Amino-4,4,4-trifluorobutyric acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L12239-03 L12239-MD
CAAAAL12239-03EA 194.34 CAD
CAAAAL12239-03 CAAAAL12239-MD
(±)-3-Amino-4,4,4-trifluorobutyric acid 97%
(±)-3-Amino-4,4,4-trifluorobutyric acid

Purity: 97% CAS No.: 584-20-3 Molecular Formula: C4H6F3NO2 Formula Weight: 157.09 Melting Point: ca 184° dec. Beilstein Reference: 1764276

Formula: C₄H₆F₃NO₂
Melting Pt: ca 184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00041413
CAS Number: 584-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR