Propanephosphonic acid cyclic anhydride ≥50% in ethyl acetate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide
L11911-09
L11911-18
CAAAAL11911-09EA
77.26
CAD
CAAAAL11911-09
CAAAAL11911-18
Propanephosphonic acid cyclic anhydride ≥50% in ethyl acetate
Propanephosphonic acid cyclic anhydride
CAS No.: 68957-94-8
Molecular Formula: C9H21O6P3
Formula Weight: 318.19
Flash Point: -3°(26°F)
Refractive Index: 1.4190
Boiling Point: 77°
UN No.: 2924
MDL No.: MFCD00006583
Formula:
C₉H₂₁O₆P₃ MW: 318.19 Boiling Pt: 77 °C Density: 1.080 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006583 CAS Number: 68957-94-8 EINECS: 422-210-5 UN: 2924 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...