N,N-Dimethylglycine methyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

L11897-06 L11897-14
CAAAAL11897-06EA 94.48 CAD
CAAAAL11897-06 CAAAAL11897-14
N,N-Dimethylglycine methyl ester 98%
N,N-Dimethylglycine methyl ester

Purity: 98% CAS No.: 7148-06-3 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Flash Point: 38°(100°F) Refractive Index: 1.4120 Boiling Point: 134-135° Beilstein Reference: 1744788 UN No.: 3272 MDL No.: MFCD00059303

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 134…135 °C
Density: 0.950
Flash Pt: 38 °C (100 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00059303
CAS Number: 7148-06-3
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR