(S)-(-)-2-Pyrrolidinone-5-carboxylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11711-14
L11711-22
L11711-36
CAAAAL11711-14EA
40.61
CAD
CAAAAL11711-14
CAAAAL11711-22
CAAAAL11711-36
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid 98+%
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid
Fieser: 1,977
Purity: 98+%
CAS No.: 98-79-3
Molecular Formula: C5H7NO3
Formula Weight: 129.12
Melting Point: 158-162°
Beilstein Reference: 82132
Merck Reference: 14,8001
Optical Rotation: -10.5° (c=5 in water)
MDL No.: MFCD00005272
Formula:
C₅H₇NO₃ Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005272 CAS Number: 98-79-3 EINECS: 202-700-3 Merck Index: 14,8001 |
Specification Test Results
Optical rotation | -10.5° (c=5 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...