(±)-Allyl-2,3-epoxypropyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11634-22
L11634-36
CAAAAL11634-22EA
66.52
CAD
CAAAAL11634-22
CAAAAL11634-36
(±)-Allyl-2,3-epoxypropyl ether 97%
(±)-Allyl 2,3-epoxypropyl ether
Purity: 97%
CAS No.: 106-92-3
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 47°(134°F)
Refractive Index: 1.4350
Odor: Ether-like
Melting Point: -100°
Boiling Point: 50-51°/15mm
Beilstein Reference: 105871
UN No.: 2219
MDL No.: MFCD00005143
Formula:
C₆H₁₀O₂ Boiling Pt: 50…51 °C/15 mm Melting Pt: –100 °C Density: 0.967 Flash Pt: 47 °C(134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005143 CAS Number: 106-92-3 EINECS: 203-442-4 UN: 2219 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...