N-(2-Pyridyl)thiourea 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11465-03
L11465-06
CAAAAL11465-03EA
78.37
CAD
CAAAAL11465-03
CAAAAL11465-06
N-(2-Pyridyl)thiourea 98%
N-(2-Pyridyl)thiourea
Purity: 98%
CAS No.: 14294-11-2
Molecular Formula: C6H7N3S
Formula Weight: 153.21
Odor: Odorless
Melting Point: 146-149°
Beilstein Reference: 117849
UN No.: 2811
MDL No.: MFCD00041227
Formula:
C₆H₇N₃S Melting Pt: 146…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041227 CAS Number: 14294-11-2 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...