Sodium methanesulfinate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11360-03
L11360-06
L11360-14
CAAAAL11360-03EA
50.88
CAD
CAAAAL11360-03
CAAAAL11360-06
CAAAAL11360-14
Sodium methanesulfinate 95%
Sodium methanesulphinate
Purity: 95%
CAS No.: 20277-69-4
Molecular Formula: CH3NaO2S
Formula Weight: 102.09
Melting Point: 227-230°
Beilstein Reference: 3565430
Formula:
CH₃NaO₂S Melting Pt: 227…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040392 CAS Number: 20277-69-4 EINECS: 243-669-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...