γ-Cyclodextrine hydrate
Supplier: Thermo Scientific Chemicals
L11271-MC
L11271-ME
CAAAAL11271-MCEA
79.32
CAD
CAAAAL11271-MC
CAAAAL11271-ME
γ-Cyclodextrine hydrate
γ-Cyclodextrine hydrate
Fieser: 9,129
CAS No.: 91464-90-3
Molecular Formula: C48H80O40.xH2O
Formula Weight: 1297.15(anhy)
Melting Point: ca 267° dec.
Beilstein Reference: 5725162
Merck Reference: 14,2718
Optical Rotation: +177° (c=1 in water) on dry basis
MDL No.: MFCD00149574
Formula:
C₄₈H₈₀O₄₀·H₂O Melting Pt: ca 267 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149574 CAS Number: 91464-90-3 EINECS: 241-482-4 Merck Index: 14,2718 |
Specification Test Results
Optical rotation | +177° (c=1 in water) on dry basis |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...