(±)-2,3-Epoxypropyl methacrylate 97% Stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Oxiran-2-ylmethyl methacrylate, Glycidyl methacrylate
Stabilization:
100 ppm Hydrochinonmonomethylether
L11133-18
L11133-30
CAAAAL11133-18EA
55.93
CAD
CAAAAL11133-18
CAAAAL11133-30
(±)-2,3-Epoxypropyl methacrylate 97% Stabilised
(±)-2,3-Epoxypropyl methacrylate
Purity: 97%
CAS No.: 106-91-2
Molecular Formula: C7H10O3
Formula Weight: 142.16
Flash Point: 83°(181°F)
Refractive Index: 1.4500
Melting Point: -82°
Boiling Point: 189°
Beilstein Reference: 2506
UN No.: 2810
MDL No.: MFCD00005137
Formula:
C₇H₁₀O₃ MW: 142.15 g/mol Boiling Pt: 189 °C Melting Pt: –82 °C Density: 1.078 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005137 CAS Number: 106-91-2 EINECS: 203-441-9 UN: 2922 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...