2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt 97%
Supplier: Thermo Scientific Chemicals
L11105-06
L11105-14
CAAAAL11105-06EA
45.03
CAD
CAAAAL11105-06
CAAAAL11105-14
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt 97%
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt
Purity: 97%
CAS No.: 4628-94-8
Molecular Formula: C2K2N2S3
Formula Weight: 226.43
Melting Point: ca 274° dec.
Beilstein Reference: 4917786
Formula:
C₂K₂N₂S₃ Melting Pt: ca 274 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003104 CAS Number: 4628-94-8 EINECS: 225-043-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...