4-Bromophenylurea 97%
Supplier: Thermo Scientific Chemicals
L11074-03
L11074-06
CAAAAL11074-03EA
49.93
CAD
CAAAAL11074-03
CAAAAL11074-06
4-Bromophenylurea 97%
4-Bromophenylurea
Purity: 97%
CAS No.: 1967-25-5
Molecular Formula: C7H7BrN2O
Formula Weight: 215.06
Beilstein Reference: 2090140
MDL No.: MFCD00025428
Formula:
C₇H₇BrN₂O Storage Temperature: Ambient |
MDL Number:
MFCD00025428 CAS Number: 1967-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...