(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) 98+%
Supplier: Thermo Scientific Chemicals
L10939-03
L10939-06
CAAAAL10939-03EA
69.2
CAD
CAAAAL10939-03
CAAAAL10939-06
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) 98+%
(2-Nitrophenyl) β-D-galactopyranoside
Purity: 98+%
CAS No.: 369-07-3
Molecular Formula: C12H15NO8
Formula Weight: 301.25
Storage Temperature: 2°C to 8°C
Melting Point: ca 192° dec.
Beilstein Reference: 92207
Optical Rotation: -68° (c=1 in water)
MDL No.: MFCD00063255
Formula:
C₁₂H₁₅NO₈ Melting Pt: ca 192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063255 CAS Number: 369-07-3 EINECS: 206-716-1 |
Specification Test Results
Optical rotation | -68° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...