Hexafluoroacetone trihydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10777-06
L10777-14
L10777-22
CAAAAL10777-06EA
57.99
CAD
CAAAAL10777-06
CAAAAL10777-14
CAAAAL10777-22
Hexafluoroacetone trihydrate 98%
Hexafluoroacetone trihydrate
Purity: 98%
CAS No.: 34202-69-2
Molecular Formula: C3F6O.3H2O
Formula Weight: 220.07 (166.02anhy)
Refractive Index: 1.3190
Melting Point: 18-21°
Boiling Point: 105-106°
Beilstein Reference: 607236
UN No.: 2552
MDL No.: MFCD00149061
Formula:
C₃F₆O·3H₂O Boiling Pt: 105…106 °C Melting Pt: 18…21 °C Density: 1.579 Storage Temperature: Ambient |
MDL Number:
MFCD00149061 CAS Number: 34202-69-2 EINECS: 211-676-3 UN: 2552 ADR: "6.1",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...