Diisopropyl azodicarboxylate 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diisopropyl diazene-1,2-dicarboxylate
L10386-14
L10386-22
CAAAAL10386-14EA
95.59
CAD
CAAAAL10386-14
CAAAAL10386-22
Diisopropyl azodicarboxylate 94%
Diisopropyl azodicarboxylate
Fieser: 14,336 15,352 17,390 21,464
Purity: 94%
CAS No.: 2446-83-5
Molecular Formula: (CH3)2CHO2C-N=N-CO2CH(CH3)2
Formula Weight: 202.21
Storage Temperature: 2°C to 8°C
Flash Point: 160°(320°F)
Refractive Index: 1.4200
Odor: Pungent
Melting Point: 3-5°
Boiling Point: 74-75°/0.3mm
Beilstein Reference: 1912326
UN No.: 3082
MDL No.: MFCD00008875
Formula:
C₈H₁₄N₂O₄ Boiling Pt: 74…75 °C/0.3 mm Melting Pt: 3…5 °C Density: 1.027 Flash Pt: 160 °C(320 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008875 CAS Number: 2446-83-5 EINECS: 219-502-8 UN: 3082 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...