Bis(2-ethylhexyl)adipate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
L10195-14
L10195-22
CAAAAL10195-14EA
78.68
CAD
CAAAAL10195-14
CAAAAL10195-22
Bis(2-ethylhexyl)adipate 99%
Bis(2-ethylhexyl) adipate
Purity: 99%
CAS No.: 103-23-1
Molecular Formula: C22H42O4
Formula Weight: 370.58
Flash Point: 196°(384°F)
Refractive Index: 1.4470
Melting Point: -67°
Boiling Point: 166-168°/1mm
Beilstein Reference: 1803774
MDL No.: MFCD00009496
Formula:
C₂₂H₄₂O₄ MW: 370.57 g/mol Boiling Pt: 166…168 °C (1 mmHg) Melting Pt: –67 °C Density: 0.990 Flash Pt: 196 °C (384 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009496 CAS Number: 103-23-1 EINECS: 203-090-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...