1-Propyl-2-thiourea 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(n-Propyl)thiourea, 1-Propylthiourea

L10149-03 L10149-06
CAAAAL10149-03EA 97.96 CAD
CAAAAL10149-03 CAAAAL10149-06
1-Propyl-2-thiourea 98%
1-Propyl-2-thiourea

Purity: 98% CAS No.: 927-67-3 Molecular Formula: C4H10N2S Formula Weight: 118.20 Odor: Odorless Melting Point: 109-112° Beilstein Reference: 1743064 UN No.: 2811 MDL No.: MFCD00041195

Formula: C₄H₁₀N₂S
MW: 118.2 g/mol
Melting Pt: 109…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00041195
CAS Number: 927-67-3
EINECS: 213-158-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR