Methyl-4,4,4-trifluoroacetoacetate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10140-06
L10140-14
CAAAAL10140-06EA
67.15
CAD
CAAAAL10140-06
CAAAAL10140-14
Methyl-4,4,4-trifluoroacetoacetate 95%
Methyl 4,4,4-trifluoroacetoacetate
Purity: 95%
CAS No.: 83643-84-9
Molecular Formula: C5H5F3O3
Formula Weight: 170.09
Flash Point: 40°(104°F)
Refractive Index: 1.3720
Boiling Point: 117-118°
Beilstein Reference: 1777343
UN No.: 3272
MDL No.: MFCD00041004
Formula:
C₅H₅F₃O₃ Boiling Pt: 117…118 °C Density: 1.340 Flash Pt: 40 °C(104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041004 CAS Number: 83643-84-9 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...