(S)-(+)-Prolinol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol
L09779-03
L09779-06
L09779-14
CAAAAL09779-03EA
50.09
CAD
CAAAAL09779-03
CAAAAL09779-06
CAAAAL09779-14
(S)-(+)-Prolinol 98%
(S)-(+)-Prolinol
Fieser: 10,332 13,261
Purity: 98%
CAS No.: 23356-96-9
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 86°(186°F)
Refractive Index: 1.4853
Melting Point: 42-44°
Boiling Point: 74-76°/2mm
Beilstein Reference: 79843
Optical Rotation: +31° (c=1 in toluene)
MDL No.: MFCD00005255
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 74…76 °C (2 mmHg) Melting Pt: 42…44 °C Density: 1.025 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005255 CAS Number: 23356-96-9 EINECS: 245-605-2 |
Specification Test Results
Optical rotation | +31° (c=1 in toluene) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...