1-Butyl-2-thiourea 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Butylthiourea
L09654-04
L09654-09
CAAAAL09654-04EA
102.23
CAD
CAAAAL09654-04
CAAAAL09654-09
1-Butyl-2-thiourea 98%
1-Butyl-2-thiourea
Purity: 98%
CAS No.: 1516-32-1
Molecular Formula: C5H12N2S
Formula Weight: 132.23
Odor: Odorless
Melting Point: 76-79°
Beilstein Reference: 1744779
MDL No.: MFCD00022173
Formula:
C₅H₁₂N₂S MW: 132.23 g/mol Melting Pt: 76…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022173 CAS Number: 1516-32-1 EINECS: 216-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...