Ethyl-3,3-diethoxypropionate 96%
Supplier: Thermo Scientific Chemicals
Warning
|
L09562-09
L09562-18
CAAAAL09562-09EA
87.06
CAD
CAAAAL09562-09
CAAAAL09562-18
Ethyl-3,3-diethoxypropionate 96%
Ethyl 3,3-diethoxypropionate
Purity: 96%
CAS No.: 10601-80-6
Molecular Formula: C9H18O4
Formula Weight: 190.24
Storage Temperature: -30°C to -10°C
Flash Point: 93°(199°F)
Refractive Index: 1.4120
Boiling Point: 92-93°/13mm
Beilstein Reference: 1772110
MDL No.: MFCD00009865
Formula:
C₉H₁₈O₄ Boiling Pt: 92…93 °C (13 mmHg) Density: 0.978 Flash Pt: 93 °C (199 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00009865 CAS Number: 10601-80-6 EINECS: 234-223-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...