(±)-2-Pyrrolidinone-5-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

L09424-18 L09424-30
CAAAAL09424-18EA 44.87 CAD
CAAAAL09424-18 CAAAAL09424-30
(±)-2-Pyrrolidinone-5-carboxylic acid 99%
(±)-2-Pyrrolidinone-5-carboxylic acid

Purity: 99% CAS No.: 149-87-1 Molecular Formula: C5H7NO3 Formula Weight: 129.12 Melting Point: 183-185° Beilstein Reference: 82131

Formula: C₅H₇NO₃
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00064322
CAS Number: 149-87-1
EINECS: 205-748-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR