Benzylthiourea 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09400-06
L09400-14
L09400-22
CAAAAL09400-06EA
61.46
CAD
CAAAAL09400-06
CAAAAL09400-14
CAAAAL09400-22
Benzylthiourea 98%
Benzylthiourea
Purity: 98%
CAS No.: 621-83-0
Molecular Formula: C8H10N2S
Formula Weight: 166.25
Odor: Odorless
Melting Point: 162-166°
Beilstein Reference: 972372
UN No.: 2811
MDL No.: MFCD00041370
Formula:
C₈H₁₀N₂S Melting Pt: 162…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041370 CAS Number: 621-83-0 EINECS: 210-709-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...