(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09031-06
L09031-14
CAAAAL09031-06EA
53.56
CAD
CAAAAL09031-06
CAAAAL09031-14
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 98+%
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione
Purity: 98+%
CAS No.: 4511-42-6
Molecular Formula: C6H8O4
Formula Weight: 144.13
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4475
Melting Point: 95-99°
Boiling Point: 255°
Beilstein Reference: 82124
Optical Rotation: +265° (c=1 in chloroform)
MDL No.: MFCD00070594
Formula:
C₆H₈O₄ Boiling Pt: 255 °C Melting Pt: 95…99 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00070594 CAS Number: 4511-42-6 EINECS: 224-832-0 |
Specification Test Results
Optical rotation | +265° (c=1 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...