3,6-Dimethyl-1,4-dioxane-2,5-dione 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DL-Lactide
L09026-09
L09026-18
CAAAAL09026-09EA
66.36
CAD
CAAAAL09026-09
CAAAAL09026-18
3,6-Dimethyl-1,4-dioxane-2,5-dione 99%
3,6-Dimethyl-1,4-dioxane-2,5-dione
Purity: 99%
CAS No.: 95-96-5
Molecular Formula: C6H8O4
Formula Weight: 144.13
Storage Temperature: 2°C to 8°C
Flash Point: 180°(356°F)
Melting Point: 122-126°
Boiling Point: 138-142°/8mm
Beilstein Reference: 82121
MDL No.: MFCD00011685
Formula:
C₆H₈O₄ MW: 144.13 g/mol Boiling Pt: 138…142 °C (8 mmHg) Melting Pt: 122…126 °C Flash Pt: 180 °C (356 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011685 CAS Number: 95-96-5 EINECS: 202-468-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...