Ethyl isobutyrylacetate 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09016-06
L09016-14
CAAAAL09016-06EA
41.87
CAD
CAAAAL09016-06
CAAAAL09016-14
Ethyl isobutyrylacetate 94%
Ethyl isobutyrylacetate
Purity: 94%
CAS No.: 7152-15-0
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 53°(127°F)
Refractive Index: 1.4265
Melting Point: -9°
Boiling Point: 172-174°
Beilstein Reference: 636726
UN No.: 3272
MDL No.: MFCD00009198
Formula:
C₈H₁₄O₃ Boiling Pt: 172…174 °C Melting Pt: –9 °C Density: 0.980 Flash Pt: 53 °C(127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009198 CAS Number: 7152-15-0 EINECS: 230-491-9 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...