Boc-D-Pro-OH 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline
L08826-03
L08826-06
CAAAAL08826-03EA
101.75
CAD
CAAAAL08826-03
CAAAAL08826-06
Boc-D-Pro-OH 98+%
Boc-D-Pro-OH
Purity: 98+%
CAS No.: 37784-17-1
Molecular Formula: C10H17NO4
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Melting Point: 133-135°
Beilstein Reference: 479316
Optical Rotation: +61° (c=1 in acetic acid)
MDL No.: MFCD00063226
Formula:
C₁₀H₁₇NO₄ MW: 215.25 g/mol Melting Pt: 133…135 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063226 CAS Number: 37784-17-1 |
Specification Test Results
Optical rotation | +61° (c=1 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...