Sodium bis(trimethylsilyl)amide 1 M in THF
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08779-AE
L08779-AP
CAAAAL08779-AEEA
92.59
CAD
CAAAAL08779-AE
CAAAAL08779-AP
Sodium bis(trimethylsilyl)amide 1 M in THF
Sodium bis(trimethylsilyl)amide
Fieser: 1,1046 3,261 4,442 6,529 7,329 12,441 18,332 20,349 21,401
CAS No.: 1070-89-9
Molecular Formula: C6H18NNaSi2
Formula Weight: 183.38
Flash Point: -17°(1°F)
Beilstein Reference: 3629917
UN No.: 2924
MDL No.: MFCD00009835
Formula:
C₆H₁₈NNaSi₂ Density: 0.904 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009835 CAS Number: 1070-89-9 EINECS: 213-983-8 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...