(4-Chlorophenylthio)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-((4-Chlorophenyl)thio)acetic acid
L08583-09
L08583-18
CAAAAL08583-09EA
100.17
CAD
CAAAAL08583-09
CAAAAL08583-18
(4-Chlorophenylthio)acetic acid 98%
(4-Chlorophenylthio)acetic acid
Purity: 98%
CAS No.: 3405-88-7
Molecular Formula: C8H7ClO2S
Formula Weight: 202.66
Melting Point: 104-107°
Beilstein Reference: 1871569
Formula:
C₈H₇ClO₂S Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021762 CAS Number: 3405-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...