Sarcosine methyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Sar-OMe·HCl, N-Sar-OMe·HCl, Methyl N-methylglycinate hydrochloride

L08572-09 L08572-18
CAAAAL08572-09EA 78.05 CAD
CAAAAL08572-09 CAAAAL08572-18
Sarcosine methyl ester hydrochloride 98%
Sarcosine methyl ester hydrochloride

Purity: 98% CAS No.: 13515-93-0 Molecular Formula: CH3NHCH2CO2CH3.HCl Formula Weight: 139.58 Melting Point: ca 105° dec. Beilstein Reference: 3677128

Formula: C₄H₉NO₂·HCl
MW: 139.58 g/mol
Melting Pt: ∼ 105 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038876
CAS Number: 13515-93-0
EINECS: 236-853-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR