2,2-Diphenylacetamide 98%

Supplier: Thermo Scientific Chemicals
Warning

L08485-06 L08485-14
CAAAAL08485-06EA 119.92 CAD
CAAAAL08485-06 CAAAAL08485-14
2,2-Diphenylacetamide 98%
2,2-Diphenylacetamide

Purity: 98% CAS No.: 4695-13-0 Molecular Formula: C14H13NO Formula Weight: 211.26 Melting Point: 164-167° Beilstein Reference: 1871583 Merck Reference: 14,3316 MDL No.: MFCD00025496

Formula: C₁₄H₁₃NO
MW: 211.26 g/mol
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00025496
CAS Number: 4695-13-0
Merck Index: 14,3316

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR