(S)-(-)-1,1'-Bi-2-naphthol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
L08423-03
L08423-06
L08423-14
CAAAAL08423-03EA
107.76
CAD
CAAAAL08423-03
CAAAAL08423-06
CAAAAL08423-14
(S)-(-)-1,1'-Bi-2-naphthol 99%
(S)-(-)-1,1'-Bi-2-naphthol
Fieser: 12,190 15,26,135 18,41 19,23 20,24 21,26
Purity: 99%
CAS No.: 18531-99-2
Molecular Formula: C20H14O2
Formula Weight: 286.33
Odor: Characteristic
Melting Point: 206-211°
Beilstein Reference: 3592999
Merck Reference: 14,1226
Optical Rotation: -34.5 (c=1 in THF)
UN No.: 2811
MDL No.: MFCD00004068
Formula:
C₂₀H₁₄O₂ Melting Pt: 206…211 °C Density: 1.301 Storage Temperature: Ambient |
MDL Number:
MFCD00004068 CAS Number: 18531-99-2 ADR: 6.1,III Merck Index: 14,1226 |
Specification Test Results
Optical rotation | -34.5° (c=1 in THF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...