6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3,4-Tetrahydro-6,7-dimethoxyquinazoline-2,4-dione, 6,7-Dimethoxyquinazoline-2,4-dione

L08411-09 L08411-18
CAAAAL08411-09EA 92.11 CAD
CAAAAL08411-09 CAAAAL08411-18
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione 98%
6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione

Purity: 98% CAS No.: 28888-44-0 Molecular Formula: C10H10N2O4 Formula Weight: 222.20 Melting Point: 300° Beilstein Reference: 204597

Formula: C₁₀H₁₀N₂O₄
MW: 222.2 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00023889
CAS Number: 28888-44-0
EINECS: 249-288-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR