Triethyl methanetricarboxylate 98%
Supplier: Thermo Scientific Chemicals
L08356-06
L08356-14
CAAAAL08356-06EA
46.61
CAD
CAAAAL08356-06
CAAAAL08356-14
Triethyl methanetricarboxylate 98%
Triethyl methanetricarboxylate
Purity: 98%
CAS No.: 6279-86-3
Molecular Formula: C10H16O6
Formula Weight: 232.23
Flash Point:
Formula:
C₁₀H₁₆O₆ MW: 232.23 g/mol Boiling Pt: 253 °C Melting Pt: 26…29 °C Density: 1,095 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009150 CAS Number: 6279-86-3 EINECS: 228-477-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...