2-Butanone oxime 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08196-AE
L08196-AP
L08196-AU
CAAAAL08196-AEEA
51.19
CAD
CAAAAL08196-AE
CAAAAL08196-AP
CAAAAL08196-AU
2-Butanone oxime 99%
2-Butanone oxime
Purity: 99%
CAS No.: 96-29-7
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point: 62°(144°F)
Refractive Index: 1.4430
Melting Point: -30°
Boiling Point: 152-153°
Beilstein Reference: 1698241
MDL No.: MFCD00013935
Formula:
C₄H₉NO Boiling Pt: 152…153 °C Melting Pt: –30 °C Density: 0.921 Flash Pt: 62 °C(144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013935 CAS Number: 96-29-7 EINECS: 202-496-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...