(±)-Epoxystyrene 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Styrene oxide

L07821-22 L07821-36
CAAAAL07821-22EA 66.04 CAD
CAAAAL07821-22 CAAAAL07821-36
(±)-Epoxystyrene 98+%
(±)-Epoxystyrene

Purity: 98+% CAS No.: 96-09-3 Molecular Formula: C8H8O Formula Weight: 120.15 Storage Temperature: 2°C to 8°C Flash Point: 74°(165°F) Refractive Index: 1.5350 Melting Point: -37° Boiling Point: 193-196° Beilstein Reference: 108582 UN No.: 2810 MDL No.: MFCD00005121

Formula: C₈H₈O
Boiling Pt: 193…196 °C
Melting Pt: –37 °C
Density: 1.053
Flash Pt: 74 °C(165 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005121
CAS Number: 96-09-3
EINECS: 202-476-7
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR