4-(2-Thienyl)butyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Thiophen-2-yl)butanoic acid

L07563-03 L07563-06
CAAAAL07563-03EA 43.92 CAD
CAAAAL07563-03 CAAAAL07563-06
4-(2-Thienyl)butyric acid 98%
4-(2-Thienyl)butyric acid

Purity: 98% CAS No.: 4653-11-6 Molecular Formula: C8H10O2S Formula Weight: 170.23 Flash Point: 110°(230°F) Refractive Index: 1.5320 Boiling Point: 122°/0.3mm Beilstein Reference: 127982 MDL No.: MFCD00005463

Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
Boiling Pt: 122 °C (0.3 mmHg)
Density: 1.169
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005463
CAS Number: 4653-11-6
EINECS: 225-090-0
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR