2-Deoxy-D-glucose 98%
Supplier: Thermo Scientific Chemicals
L07338-03
L07338-06
L07338-14
CAAAAL07338-03EA
106.02
CAD
CAAAAL07338-03
CAAAAL07338-06
CAAAAL07338-14
2-Deoxy-D-glucose 98%
2-Deoxy-D-glucose
A useful culture media component for molecular genetics
Purity: 98%
CAS No.: 154-17-6
Molecular Formula: C6H12O5
Formula Weight: 164.16
Melting Point: 147-152°
Beilstein Reference: 1723331
Merck Reference: 14,2904
Optical Rotation: +47° (c=0.5 in water, 45h)
MDL No.: MFCD00151328
Formula:
C₆H₁₂O₅ Melting Pt: 147…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151328 CAS Number: 154-17-6 EINECS: 205-823-0 Merck Index: 14,2904 |
Specification Test Results
Optical rotation | +47° (c=0.5 in water, 45h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...