1,2-Epoxydecane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07289-06
L07289-14
CAAAAL07289-06EA
57.51
CAD
CAAAAL07289-06
CAAAAL07289-14
1,2-Epoxydecane 97%
1,2-Epoxydecane
Purity: 97%
CAS No.: 2404-44-6
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 78°(172°F)
Refractive Index: 1.4290
Boiling Point: 92-93°/14mm
Beilstein Reference: 104132
MDL No.: MFCD00005158
Formula:
C₁₀H₂₀O Boiling Pt: 92…93 °C/14 mm Density: 0,840 Flash Pt: 78 °C(172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005158 CAS Number: 2404-44-6 EINECS: 219-295-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...