2-Amino-5-tert-butyl-1,3,4-thiadiazole 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-(tert-Butyl)-1,3,4-thiadiazol-2-amine
L06116-03
L06116-06
CAAAAL06116-03EA
37.13
CAD
CAAAAL06116-03
CAAAAL06116-06
2-Amino-5-tert-butyl-1,3,4-thiadiazole 97%
2-Amino-5-tert-butyl-1,3,4-thiadiazole
Purity: 97%
CAS No.: 39222-73-6
Molecular Formula: C6H11N3S
Formula Weight: 157.24
Melting Point: 181-185°
Beilstein Reference: 119853
Formula:
C₆H₁₁N₃S Melting Pt: 181…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040403 CAS Number: 39222-73-6 EINECS: 254-364-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...