6,9-Dichloro-2-methoxyacridine 98%

Supplier: Thermo Scientific Chemicals
Danger

L05913-06 L05913-14
CAAAAL05913-06EA 65.25 CAD
CAAAAL05913-06 CAAAAL05913-14
6,9-Dichloro-2-methoxyacridine 98%
6,9-Dichloro-2-methoxyacridine

Purity: 98% CAS No.: 86-38-4 Molecular Formula: C14H9Cl2NO Formula Weight: 278.14 Melting Point: 163-165° Beilstein Reference: 226309

Formula: C₁₄H₉Cl₂NO
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00005028
CAS Number: 86-38-4
EINECS: 201-666-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR